7-Fluoroindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS12775G1 AS12775G25 AS12775G5
ACTIAS12775G1EA 0 GBP
ACTIAS12775G1 ACTIAS12775G25 ACTIAS12775G5
7-Fluoroindole
7-Fluoroindole
Formula: C₈H₆FN
MDL Number: MFCD01074502
CAS Number: 387-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR