7-Bromoindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS51137G1 AS51137G25 AS51137G5
ACTIAS51137G1EA 0 GBP
ACTIAS51137G1 ACTIAS51137G25 ACTIAS51137G5
7-Bromoindole
7-Bromoindole
Formula: C₈H₆BrN
MDL Number: MFCD00799492
CAS Number: 51417-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR