6-Nitroindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Nitro-1H-indole

AS21033G25 AS21033G5
ACTIAS21033G25EA 97 GBP
ACTIAS21033G25 ACTIAS21033G5
6-Nitroindole
6-Nitroindole
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00051497
CAS Number: 4769-96-4
EINECS: 225-311-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR