6-Hydroxyindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS50084G1 AS50084G5
ACTIAS50084G1EA 0 GBP
ACTIAS50084G1 ACTIAS50084G5
6-Hydroxyindole
6-Hydroxyindole
Formula: C₈H₇NO
MDL Number: MFCD00152101
CAS Number: 2380-86-1
EINECS: 417-020-4
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR