5-Hydroxyindole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS50144G25
AS50144G5
ACTIAS50144G25EA
0
GBP
ACTIAS50144G25
ACTIAS50144G5
5-Hydroxyindole
5-Hydroxyindole
Formula:
C₈H₇NO Melting Pt: 107…108 °C |
MDL Number:
MFCD00005677 CAS Number: 1953-54-4 EINECS: 217-782-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...