4-Hydroxyphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20536G25 AS20536G5
ACTIAS20536G25EA 0 GBP
ACTIAS20536G25 ACTIAS20536G5
4-Hydroxyphenylboronic acid
4-Hydroxyphenylboronic acid
Formula: C₆H₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS Number: 71597-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR