4-Fluoro-m-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22668G100 AS22668G25
ACTIAS22668G100EA 0 GBP
ACTIAS22668G100 ACTIAS22668G25
4-Fluoro-m-toluic acid
4-Fluoro-m-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD01631424
CAS Number: 403-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR