4-Fluoro-o-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22260G100 AS22260G25
ACTIAS22260G100EA 0 GBP
ACTIAS22260G100 ACTIAS22260G25
4-Fluoro-o-toluic acid
4-Fluoro-o-toluic acid
Formula: C₈H₇FO₂
MDL Number: MFCD04115880
CAS Number: 321-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR