4-Cyanophenylboronic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane
AS50164G100
AS50164G25
AS50164G5
ACTIAS50164G100EA
158
GBP
ACTIAS50164G100
ACTIAS50164G25
ACTIAS50164G5
4-Cyanophenylboronic acid
4-Cyanophenylboronic acid
Formula:
C₇H₆BNO₂ MW: 146.94 g/mol Melting Pt: > 300 °C |
MDL Number:
MFCD01318968 CAS Number: 126747-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...