4-Bromo-3-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS23047G100 AS23047G25
ACTIAS23047G100EA 0 GBP
ACTIAS23047G100 ACTIAS23047G25
4-Bromo-3-fluorobenzoic acid
4-Bromo-3-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MDL Number: MFCD00672932
CAS Number: 153556-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR