4-Bromo-2-nitrobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20209G100
AS20209G25
ACTIAS20209G100EA
0
GBP
ACTIAS20209G100
ACTIAS20209G25
4-Bromo-2-nitrobenzoic acid
4-Bromo-2-nitrobenzoic acid
Formula:
C₇H₄BrNO₄ |
MDL Number:
MFCD01013599 CAS Number: 99277-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...