4-Amino-3,5-dichloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dichloro-4-pyridylamine, 3,5-Dichloropyridin-4-amine

AS24342G100 AS24342G25
ACTIAS24342G100EA 75 GBP
ACTIAS24342G100 ACTIAS24342G25
4-Amino-3,5-dichloropyridine
4-Amino-3,5-dichloropyridine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
MDL Number: MFCD00125023
CAS Number: 22889-78-7
EINECS: 245-304-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR