5-Fluoroisatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20152G100 AS20152G25
ACTIAS20152G100EA 0 GBP
ACTIAS20152G100 ACTIAS20152G25
5-Fluoroisatin
5-Fluoroisatin
Formula: C₈H₄FNO₂
Storage Temperature: Ambient
MDL Number: MFCD00022795
CAS Number: 443-69-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR