3-Iodo-p-toluic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20483G100
AS20483G25
ACTIAS20483G100EA
0
GBP
ACTIAS20483G100
ACTIAS20483G25
3-Iodo-p-toluic acid
3-Iodo-p-toluic acid
Formula:
C₈H₇IO₂ MW: 262.05 g/mol Boiling Pt: 329 Melting Pt: 210…211 °C |
MDL Number:
MFCD00010392 CAS Number: 82998-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...