3-Fluorophenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid

AS20248G100 AS20248G25
ACTIAS20248G100EA 0 GBP
ACTIAS20248G100 ACTIAS20248G25
3-Fluorophenylboronic acid
3-Fluorophenylboronic acid
Formula: C₆H₆BFO₂
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00236042
CAS Number: 768-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR