5-Chloro-2-methylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Chloro-o-toluic acid

AS20452G5 AS20452G100 AS20452G25
ACTIAS20452G5EA 0 GBP
ACTIAS20452G5 ACTIAS20452G100 ACTIAS20452G25
5-Chloro-2-methylbenzoic acid
5-Chloro-2-methylbenzoic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00045842
CAS Number: 7499-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR