5-Bromo-2-hydroxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Bromo-2-pyridinol, 3-Bromo-6-hydroxypyridine, 3-Bromo-6-pyridinol, 5-Bromopyridin-2-ol

AS20790G100 AS20790G25
ACTIAS20790G100EA 0 GBP
ACTIAS20790G100 ACTIAS20790G25
5-Bromo-2-hydroxypyridine
5-Bromo-2-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
MDL Number: MFCD00234042
CAS Number: 13466-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR