5-Bromo-2-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS23140G100 AS23140G25
ACTIAS23140G100EA 104 GBP
ACTIAS23140G100 ACTIAS23140G25
5-Bromo-2-fluorobenzoic acid
5-Bromo-2-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00143423
CAS Number: 146328-85-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR