3-Bromo-5-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS22718G25 AS22718G5
ACTIAS22718G25EA 0 GBP
ACTIAS22718G25 ACTIAS22718G5
3-Bromo-5-nitrobenzoic acid
3-Bromo-5-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MDL Number: MFCD00100098
CAS Number: 6307-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR