3-Bromo-5-hydroxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Bromo-3-pyridinol, 3-Bromo-5-pyridinol, 5-Bromo-pyridin-3-ol

AS21028G25 AS21028G5
ACTIAS21028G25EA 0 GBP
ACTIAS21028G25 ACTIAS21028G5
3-Bromo-5-hydroxypyridine
3-Bromo-5-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
MDL Number: MFCD00661305
CAS Number: 74115-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR