3-Bromo-2-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22769G1 AS22769G25 AS22769G5
ACTIAS22769G1EA 0 GBP
ACTIAS22769G1 ACTIAS22769G25 ACTIAS22769G5
3-Bromo-2-methoxypyridine
3-Bromo-2-methoxypyridine
Formula: C₆H₆BrNO
MDL Number: MFCD03095349
CAS Number: 13472-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR