3-Bromo-2-fluoropyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS51141G1
AS51141G25
AS51141G5
ACTIAS51141G1EA
0
GBP
ACTIAS51141G1
ACTIAS51141G25
ACTIAS51141G5
3-Bromo-2-fluoropyridine
3-Bromo-2-fluoropyridine
Formula:
C₅H₃BrFN |
MDL Number:
MFCD04112496 CAS Number: 36178-05-9 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...