3-Bromo-2-fluorobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20607G100
AS20607G25
AS20607G5
ACTIAS20607G100EA
0
GBP
ACTIAS20607G100
ACTIAS20607G25
ACTIAS20607G5
3-Bromo-2-fluorobenzoic acid
3-Bromo-2-fluorobenzoic acid
Formula:
C₇H₄BrFO₂ MW: 219.01 g/mol |
MDL Number:
MFCD00665763 CAS Number: 161957-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...