3-Amino-4-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21323G100 AS21323G25
ACTIAS21323G100EA 0 GBP
ACTIAS21323G100 ACTIAS21323G25
3-Amino-4-methylpyridine
3-Amino-4-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00128871
CAS Number: 3430-27-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR