3-Amino-2-nitropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21045G25 AS21045G5
ACTIAS21045G25EA 0 GBP
ACTIAS21045G25 ACTIAS21045G5
3-Amino-2-nitropyridine
3-Amino-2-nitropyridine
Formula: C₅H₅N₃O₂
Melting Pt: 203…204 °C
MDL Number: MFCD00044102
CAS Number: 13269-19-7
EINECS: 236-260-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR