3,5-Dimethylphenylboronic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid
AS21730G25
AS21730G5
ACTIAS21730G25EA
59
GBP
ACTIAS21730G25
ACTIAS21730G5
3,5-Dimethylphenylboronic acid
3,5-Dimethylphenylboronic acid
Formula:
C₈H₁₁BO₂ MW: 149.98 g/mol |
MDL Number:
MFCD00185689 CAS Number: 172975-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...