3,4,5-Trichloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS24341G100 AS24341G25 AS24341G5
ACTIAS24341G100EA 0 GBP
ACTIAS24341G100 ACTIAS24341G25 ACTIAS24341G5
3,4,5-Trichloropyridine
3,4,5-Trichloropyridine
Formula: C₅H₂Cl₃N
MW: 182.44 g/mol
MDL Number: MFCD00051685
CAS Number: 33216-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR