6-Bromo-2-hydroxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-2-pyridone, 2-Bromo-6-hydroxypyridine, 6-Bromo-2(1H)-pyridinone, 6-Bromo-2-pyridinol

AS25045G1 AS25045G5
ACTIAS25045G1EA 0 GBP
ACTIAS25045G1 ACTIAS25045G5
6-Bromo-2-hydroxypyridine
6-Bromo-2-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
MDL Number: MFCD00234043
CAS Number: 27992-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR