2-Bromo-6-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21311G25 AS21311G5
ACTIAS21311G25EA 0 GBP
ACTIAS21311G25 ACTIAS21311G5
2-Bromo-6-fluorobenzoic acid
2-Bromo-6-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MDL Number: MFCD01569539
CAS Number: 2252-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR