2,3-Difluorophenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21838G100 AS21838G25 AS21838G5
ACTIAS21838G100EA 0 GBP
ACTIAS21838G100 ACTIAS21838G25 ACTIAS21838G5
2,3-Difluorophenylboronic acid
2,3-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MDL Number: MFCD01863170
CAS Number: 121219-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR