1-Iodo-4,4,4-trifluorobutane
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Iodo-1,1,1-trifluorobutane, 1,1,1-Trifluoro-4-iodobutane
AS23930G25
AS23930G5
ACTIAS23930G25EA
0
GBP
ACTIAS23930G25
ACTIAS23930G5
1-Iodo-4,4,4-trifluorobutane
1-Iodo-4,4,4-trifluorobutane
Formula:
C₄H₆F₃I MW: 237.99 g/mol |
MDL Number:
MFCD00041543 CAS Number: 461-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...