3-Nitro-p-cresol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21037G100 AS21037G25
ACTIAS21037G100EA 0 GBP
ACTIAS21037G100 ACTIAS21037G25
3-Nitro-p-cresol
3-Nitro-p-cresol
Formula: C₇H₇NO₃
Melting Pt: 78…81 °C
MDL Number: MFCD00007244
CAS Number: 2042-14-0
EINECS: 218-044-6
UN: 2446
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR