2-Nitro-m-cresol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21652G100
AS21652G25
ACTIAS21652G100EA
0
GBP
ACTIAS21652G100
ACTIAS21652G25
2-Nitro-m-cresol
2-Nitro-m-cresol
Formula:
C₇H₇NO₃ Boiling Pt: 231 °C (1013 hPa) Melting Pt: 35…39 °C |
MDL Number:
MFCD00007125 CAS Number: 4920-77-8 EINECS: 225-546-9 UN: 2446 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...