3-Nitro-o-cresol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20903G100 AS20903G25
ACTIAS20903G100EA 0 GBP
ACTIAS20903G100 ACTIAS20903G25
3-Nitro-o-cresol
3-Nitro-o-cresol
Formula: C₇H₇NO₃
Melting Pt: 149…152 °C
MDL Number: MFCD00007241
CAS Number: 5460-31-1
EINECS: 226-739-0
UN: 2446
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR