4-Fluoro-2-nitroanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS24935G100 AS24935G25
ACTIAS24935G100EA 0 GBP
ACTIAS24935G100 ACTIAS24935G25
4-Fluoro-2-nitroanisole
4-Fluoro-2-nitroanisole
Formula: C₇H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD00013375
CAS Number: 445-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR