5-Fluoro-o-cresol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Fluoro-2-methylphenol, 1-Fluoro-3-hydroxy-4-methylbenzene

AS21034G100 AS21034G25
ACTIAS21034G100EA 0 GBP
ACTIAS21034G100 ACTIAS21034G25
5-Fluoro-o-cresol
5-Fluoro-o-cresol
Formula: C₇H₇FO
MDL Number: MFCD00673009
CAS Number: 452-85-7
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR