4-Chlorobenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Chlorophenyl)methanol

AS20057G100 AS20057G25
ACTIAS20057G100EA 0 GBP
ACTIAS20057G100 ACTIAS20057G25
4-Chlorobenzyl alcohol
4-Chlorobenzyl alcohol
Formula: C₇H₇ClO
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 67…70 °C
MDL Number: MFCD00004652
CAS Number: 873-76-7
EINECS: 212-852-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR