5-Chloro-o-cresol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Chloro-3-hydroxy-4-methylbenzene, 5-Chloro-2-methylphenol
AS24942G100
AS24942G25
AS24942G5
ACTIAS24942G100EA
0
GBP
ACTIAS24942G100
ACTIAS24942G25
ACTIAS24942G5
5-Chloro-o-cresol
5-Chloro-o-cresol
Formula:
C₇H₇ClO MW: 142.58 g/mol Boiling Pt: 225 Melting Pt: 75…76 °C |
MDL Number:
MFCD00060672 CAS Number: 5306-98-9 EINECS: 226-160-3 UN: 3437 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...