5-Chloro-m-xylene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24905G25
AS24905G5
ACTIAS24905G25EA
132
GBP
ACTIAS24905G25
ACTIAS24905G5
5-Chloro-m-xylene
5-Chloro-m-xylene
Formula:
C₈H₉Cl |
MDL Number:
MFCD00060273 CAS Number: 556-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...