2-Chloro-5-nitrophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20721G100 AS20721G25
ACTIAS20721G100EA 193 GBP
ACTIAS20721G100 ACTIAS20721G25
2-Chloro-5-nitrophenol
2-Chloro-5-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
MDL Number: MFCD01571825
CAS Number: 619-10-3
EINECS: 210-579-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR