2-Bromo-α,α,α-trifluoro-5-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24958G100 AS24958G25
ACTIAS24958G100EA 0 GBP
ACTIAS24958G100 ACTIAS24958G25
2-Bromo-α,α,α-trifluoro-5-nitrotoluene
2-Bromo-α,α,α-trifluoro-5-nitrotoluene
Formula: C₇H₃BrF₃NO₂
Boiling Pt: 125…130 °C (12 mmHg)
Melting Pt: 46…48 °C
MDL Number: MFCD00014707
CAS Number: 367-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR