3-Bromobenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Bromophenyl)methanol

AS21364G100 AS21364G25
ACTIAS21364G100EA 0 GBP
ACTIAS21364G100 ACTIAS21364G25
3-Bromobenzyl alcohol
3-Bromobenzyl alcohol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 165
Density: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00004629
CAS Number: 15852-73-0
EINECS: 239-975-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR