2-Bromo-3-fluorotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23527G25 AS23527G5
ACTIAS23527G25EA 0 GBP
ACTIAS23527G25 ACTIAS23527G5
2-Bromo-3-fluorotoluene
2-Bromo-3-fluorotoluene
Formula: C₇H₆BrF
MDL Number: MFCD08458010
CAS Number: 59907-13-0
EINECS: 261-982-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR