4-Bromo-3,5-difluorophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20545G1 AS20545G25 AS20545G5
ACTIAS20545G1EA 0 GBP
ACTIAS20545G1 ACTIAS20545G25 ACTIAS20545G5
4-Bromo-3,5-difluorophenol
4-Bromo-3,5-difluorophenol
Formula: C₆H₃BrF₂O
MDL Number: MFCD08062385
CAS Number: 130191-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR