4-Bromo-3-fluoroaniline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS22315G100
AS22315G25
ACTIAS22315G100EA
0
GBP
ACTIAS22315G100
ACTIAS22315G25
4-Bromo-3-fluoroaniline
4-Bromo-3-fluoroaniline
Formula:
C₆H₅BrFN Boiling Pt: 231 °C (1013 hPa) Melting Pt: 72…73 °C |
MDL Number:
MFCD00672933 CAS Number: 656-65-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...