5-Fluoro-2-nitroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22548G100 AS22548G25
ACTIAS22548G100EA 0 GBP
ACTIAS22548G100 ACTIAS22548G25
5-Fluoro-2-nitroaniline
5-Fluoro-2-nitroaniline
Formula: C₆H₅FN₂O₂
MDL Number: MFCD00034065
CAS Number: 2369-11-1
EINECS: 219-128-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR