3-Fluoro-o-toluidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21591G100
AS21591G25
ACTIAS21591G100EA
0
GBP
ACTIAS21591G100
ACTIAS21591G25
3-Fluoro-o-toluidine
3-Fluoro-o-toluidine
Formula:
C₇H₈FN Boiling Pt: 203 °C (1013 hPa) Melting Pt: 7 °C Density: 1.359 g/cm³ (20 °C) |
MDL Number:
MFCD00007760 CAS Number: 443-86-7 EINECS: 207-142-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...