3-Bromo-p-toluidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Amino-2-bromotoluene, 3-Bromo-4-methylaniline

AS21236G100 AS21236G25
ACTIAS21236G100EA 0 GBP
ACTIAS21236G100 ACTIAS21236G25
3-Bromo-p-toluidine
3-Bromo-p-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 254…257 °C (1013 hPa)
Melting Pt: 25…26 °C
MDL Number: MFCD00134176
CAS Number: 7745-91-7
EINECS: 231-807-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR