4,5-Difluoro-2-nitroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,4-Difluoro-6-nitroaniline

AS21172G100 AS21172G25
ACTIAS21172G100EA 0 GBP
ACTIAS21172G100 ACTIAS21172G25
4,5-Difluoro-2-nitroaniline
4,5-Difluoro-2-nitroaniline
Formula: C₆H₄F₂N₂O₂
MW: 174.11 g/mol
MDL Number: MFCD00010876
CAS Number: 78056-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR