4,5-Dichloro-2-nitroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS20825G100 AS20825G25
ACTIAS20825G100EA 0 GBP
ACTIAS20825G100 ACTIAS20825G25
4,5-Dichloro-2-nitroaniline
4,5-Dichloro-2-nitroaniline
Formula: C₆H₄Cl₂N₂O₂
MW: 207.02 g/mol
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007770
CAS Number: 6641-64-1
EINECS: 229-657-3
UN: 2237
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR